2-(Benzyloxy)-1-bromo-3chloro-4-fluorobenzene ≥95%

Supplier: AOBChem USA

Warning

34031-500MG 34031-1G 34031-5G 34031-25G
77325-612EA 262.65 USD
77325-612 77349-282 77361-978 77385-012
2-(Benzyloxy)-1-bromo-3chloro-4-fluorobenzene ≥95%
2-(Benzyloxy)-1-bromo-3chloro-4-fluorobenzene

Precaución: for Research Use Only

Formula: C₁₃H₉BrClFO
Storage Temperature: Ambient
MDL Number: MFCD31619457
CAS Number: 2179038-30-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR