1-(3-Bromo-4-fluoro-5-hydroxyphenyl)ethan-1-one ≥95%

Supplier: AOBChem USA

Warning

33958-500MG 33958-1G 33958-5G
77339-564EA 597.4 USD
77339-564 77349-278 77373-534
1-(3-Bromo-4-fluoro-5-hydroxyphenyl)ethan-1-one ≥95%
1-(3-Bromo-4-fluoro-5-hydroxyphenyl)ethan-1-one

Precaución: for Research Use Only

Formula: C₈H₆BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD28738810
CAS Number: 1780897-84-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR