Ethyl 4-bromo-2-chloro-6-methylbenzoate ≥95%

Supplier: AOBChem USA

Warning

33942-500MG 33942-1G 33942-25G 33942-5G
77325-610EA 284.28 USD
77325-610 77349-274 77385-010 77373-530
Ethyl 4-bromo-2-chloro-6-methylbenzoate ≥95%
Ethyl 4-bromo-2-chloro-6-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrClO₂
Storage Temperature: Ambient
MDL Number: MFCD27965165
CAS Number: 1823870-34-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR