Potassium trifluoro(2-fluoro-5-methoxyphenyl)boranuide ≥95%

Supplier: AOBChem USA

Warning

33909-500MG 33909-1G 33909-5G 33909-25G
77325-606EA 178.19 USD
77325-606 77346-640 77361-976 77385-008
Potassium trifluoro(2-fluoro-5-methoxyphenyl)boranuide ≥95%
Potassium trifluoro(2-fluoro-5-methoxyphenyl)boranuide

Precaución: for Research Use Only

Formula: C₇H₆BF₄KO
Storage Temperature: Ambient
MDL Number: MFCD09993356
CAS Number: 1985700-38-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR