6-Bromo-3-ethoxy-2-fluorophenol ≥97%

Supplier: AOBChem USA

Warning

33896-500MG 33896-1G 33896-5G 33896-25G
77325-604EA 263.68 USD
77325-604 77349-270 77361-974 77385-006
6-Bromo-3-ethoxy-2-fluorophenol ≥97%
6-Bromo-3-ethoxy-2-fluorophenol

Precaución: for Research Use Only

Formula: C₈H₈BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD28738557
CAS Number: 1776923-59-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR