Methyl 3-chloro-2-fluoroisonicotinate ≥97%

Supplier: AOBChem USA

Warning

33890-500MG 33890-1G 33890-5G 33890-25G
77325-600EA 263.68 USD
77325-600 77349-266 77361-970 77385-002
Methyl 3-chloro-2-fluoroisonicotinate ≥97%
Methyl 3-chloro-2-fluoroisonicotinate

Precaución: for Research Use Only

Formula: C₇H₅ClFNO₂
Storage Temperature: Ambient
MDL Number: MFCD12911469
CAS Number: 1214385-95-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR