3-Chloro-2-fluoro-4-(methylthio)pyridine ≥95%

Supplier: AOBChem USA

Warning

33885-500MG 33885-1G 33885-5G 33885-25G
77325-598EA 91.67 USD
77325-598 77342-424 77361-968 77373-526
3-Chloro-2-fluoro-4-(methylthio)pyridine ≥95%
3-Chloro-2-fluoro-4-(methylthio)pyridine

Precaución: for Research Use Only

Formula: C₆H₅ClFNS
MW: 177.627 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28130422
CAS Number: 1823323-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR