Ethyl 4-chloro-3-fluoro-2-methybenzoate ≥95%

Supplier: AOBChem USA

Warning

33843-500MG 33843-1G 33843-5G
77338-288EA 473.8 USD
77338-288 77349-256 77373-520
Ethyl 4-chloro-3-fluoro-2-methybenzoate ≥95%
Ethyl 4-chloro-3-fluoro-2-methybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀ClFO₂
Storage Temperature: Ambient
MDL Number: MFCD28751869
CAS Number: 1807027-04-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR