Methyl 2,6-Dichloro-4-(methylthio)benzoate ≥97%

Supplier: AOBChem USA

Warning

33835-500MG 33835-1G 33835-5G 33835-25G
77325-582EA 110.21 USD
77325-582 77343-258 77361-954 77373-514
Methyl 2,6-Dichloro-4-(methylthio)benzoate ≥97%
Methyl 2,6-Dichloro-4-(methylthio)benzoate

Precaución: for Research Use Only

Formula: C₉H₈Cl₂O₂S
MW: 251.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27987155
CAS Number: 1823359-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR