Methyl 4-chloro-2-fluoro-3-(methylthio)benzoate ≥97%

Supplier: AOBChem USA

Warning

33817-500MG 33817-1G 33817-5G 33817-25G
77325-578EA 182.31 USD
77325-578 77346-918 77361-950 77384-988
Methyl 4-chloro-2-fluoro-3-(methylthio)benzoate ≥97%
Methyl 4-chloro-2-fluoro-3-(methylthio)benzoate

Precaución: for Research Use Only

Formula: C₉H₈ClFO₂S
MW: 234.675 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31561333
CAS Number: 159329-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR