4-Methyl-5-(methylsulfanyl)pyridine-3-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

33750-500MG 33750-1G 33750-5G
77339-044EA 547.96 USD
77339-044 77349-246 77373-504
4-Methyl-5-(methylsulfanyl)pyridine-3-carboxylic acid ≥97%
4-Methyl-5-(methylsulfanyl)pyridine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₉NO₂S
MW: 183.228 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31561323
CAS Number: 2145093-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR