1-Bromo-3-(methylthio)-5-(phenylmethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

33727-500MG 33727-1G 33727-5G
77336-454EA 388.31 USD
77336-454 77349-244 77373-502
1-Bromo-3-(methylthio)-5-(phenylmethoxy)benzene ≥97%
1-Bromo-3-(methylthio)-5-(phenylmethoxy)benzene

Precaución: for Research Use Only

Formula: C₁₄H₁₃BrOS
MW: 309.221 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34755938
CAS Number: 1186604-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR