Methyl 2-fluoro-5-isopropoxynicotinate ≥95%

Supplier: AOBChem USA

Warning

33715-500MG 33715-1G 33715-5G 33715-25G
77325-570EA 208.06 USD
77325-570 77349-240 77361-944 77384-984
Methyl 2-fluoro-5-isopropoxynicotinate ≥95%
Methyl 2-fluoro-5-isopropoxynicotinate

Precaución: for Research Use Only

Formula: C₁₀H₁₂FNO₃
MW: 213.206 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31561319
CAS Number: 2145093-93-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR