Ethyl 2-bromo-3,6-difluorobenzoate ≥95%

Supplier: AOBChem USA

Warning

33710-500MG 33710-1G 33710-5G 33710-25G
77325-568EA 243.08 USD
77325-568 77349-238 77361-942 77384-982
Ethyl 2-bromo-3,6-difluorobenzoate ≥95%
Ethyl 2-bromo-3,6-difluorobenzoate

Precaución: for Research Use Only

Formula: C₉H₇BrF₂O₂
MW: 265.051 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28061435
CAS Number: 1804408-94-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR