(5-(Benzyloxy)-2-bromophenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

33707-1G 33707-5G 33707-25G
77340-874EA 61.8 USD
77340-874 77361-940 77373-498
(5-(Benzyloxy)-2-bromophenyl)methanol ≥97%
(5-(Benzyloxy)-2-bromophenyl)methanol

Precaución: for Research Use Only

Formula: C₁₄H₁₃BrO₂
MW: 293.156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24131229
CAS Number: 1084897-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR