2-Bromo-4,5-dimethylphenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

33698-500MG 33698-1G 33698-5G 33698-25G
77325-566EA 85.49 USD
77325-566 77342-242 77361-938 77373-496
2-Bromo-4,5-dimethylphenylboronic acid ≥95%
2-Bromo-4,5-dimethylphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BBrO₂
Storage Temperature: Ambient
MDL Number: MFCD31561314
CAS Number: 2138894-45-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR