Methyl 3-chloro-5-fluoro-2-isopropoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

33690-500MG 33690-1G 33690-5G 33690-25G
77325-564EA 182.31 USD
77325-564 77346-916 77361-936 77384-980
Methyl 3-chloro-5-fluoro-2-isopropoxybenzoate ≥97%
Methyl 3-chloro-5-fluoro-2-isopropoxybenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₂ClFO₃
MW: 246.663 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31561313
CAS Number: 2121777-23-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR