3-Bromo-2,5-dichlorobenzyl alcohol ≥95%

Supplier: AOBChem USA

Warning

33674-500MG 33674-1G 33674-5G
77338-550EA 498.52 USD
77338-550 77349-234 77373-494
3-Bromo-2,5-dichlorobenzyl alcohol ≥95%
3-Bromo-2,5-dichlorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₅BrCl₂O
Storage Temperature: Ambient
MDL Number: MFCD28737835
CAS Number: 1805119-74-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR