3,4-Diphenyl-1-(trifluoromethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

33664-500MG 33664-1G 33664-5G 33664-25G
77325-556EA 144.2 USD
77325-556 77345-044 77361-928 77384-974
3,4-Diphenyl-1-(trifluoromethoxy)benzene ≥97%
3,4-Diphenyl-1-(trifluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₁₉H₁₃F₃O
Storage Temperature: Ambient
MDL Number: MFCD31382565
CAS Number: 2121515-35-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR