1-Bromo-4,5-dimethyl-2-iodobenzene ≥95%

Supplier: AOBChem USA

Warning

33658-500MG 33658-1G 33658-5G 33658-25G
77325-554EA 263.68 USD
77325-554 77349-232 77361-926 77384-972
1-Bromo-4,5-dimethyl-2-iodobenzene ≥95%
1-Bromo-4,5-dimethyl-2-iodobenzene

Precaución: for Research Use Only

Formula: C₈H₈BrI
MW: 310.958 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00192708
CAS Number: 96843-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR