3-Fluoro-4-methyl-2-(methylsulfanyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

33651-500MG 33651-1G 33651-5G 33651-25G
77325-552EA 122.57 USD
77325-552 77343-984 77361-924 77373-490
3-Fluoro-4-methyl-2-(methylsulfanyl)benzoic acid ≥97%
3-Fluoro-4-methyl-2-(methylsulfanyl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₂S
Storage Temperature: Ambient
MDL Number: MFCD30538231
CAS Number: 2091775-92-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR