[3-Fluoro-4-methyl-2-(methylsulfanyl)phenyl]methanol ≥97%

Supplier: AOBChem USA

Warning

33650-500MG 33650-1G 33650-5G 33650-25G
77325-550EA 182.31 USD
77325-550 77346-914 77361-922 77384-970
[3-Fluoro-4-methyl-2-(methylsulfanyl)phenyl]methanol ≥97%
[3-Fluoro-4-methyl-2-(methylsulfanyl)phenyl]methanol

Precaución: for Research Use Only

Formula: C₉H₁₁FOS
Storage Temperature: Ambient
MDL Number: MFCD31561309
CAS Number: 2145093-92-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR