3-Fluoro-4-methyl-2-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

33649-500MG 33649-1G 33649-5G 33649-25G
77325-548EA 122.57 USD
77325-548 77343-982 77361-920 77373-488
3-Fluoro-4-methyl-2-(methylthio)benzaldehyde ≥97%
3-Fluoro-4-methyl-2-(methylthio)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉FOS
MW: 184.231 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31382564
CAS Number: 2092780-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR