2-(6-Bromo-2,3-difluorophenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

33638-500MG 33638-1G 33638-5G 33638-25G
77325-544EA 122.57 USD
77325-544 77343-980 77361-916 77373-486
2-(6-Bromo-2,3-difluorophenyl)propan-2-ol ≥95%
2-(6-Bromo-2,3-difluorophenyl)propan-2-ol

Precaución: for Research Use Only

Formula: C₉H₉BrF₂O
MW: 251.068 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30294947
CAS Number: 173276-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR