4-Ethoxy-2,5-difluorophenol ≥95%

Supplier: AOBChem USA

Warning

33626-500MG 33626-1G 33626-5G 33626-25G
77325-542EA 120.51 USD
77325-542 77343-678 77361-914 77373-484
4-Ethoxy-2,5-difluorophenol ≥95%
4-Ethoxy-2,5-difluorophenol

Precaución: for Research Use Only

Formula: C₈H₈F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD24728303
CAS Number: 612092-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR