3-(Benzyloxy)-5-bromophenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

33597-500MG 33597-1G 33597-5G 33597-25G
77325-540EA 156.56 USD
77325-540 77345-608 77361-912 77384-966
3-(Benzyloxy)-5-bromophenylboronic acid pinacol ester ≥97%
3-(Benzyloxy)-5-bromophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₉H₂₂BBrO₃
MW: 389.091 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31382561
CAS Number: 2121515-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR