2,5-Dichloro-3-methoxyphenol ≥95%

Supplier: AOBChem USA

Warning

33574-500MG 33574-1G 33574-5G 33574-25G
77325-536EA 284.28 USD
77325-536 77349-228 77361-906 77384-962
2,5-Dichloro-3-methoxyphenol ≥95%
2,5-Dichloro-3-methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₆Cl₂O₂
MW: 193.027 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24716581
CAS Number: 1781571-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR