4-Ethoxy-2-(trifluoromethyl)phenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

33563-500MG 33563-1G 33563-5G 33563-25G
77325-530EA 134.93 USD
77325-530 77344-494 77361-900 77384-956
4-Ethoxy-2-(trifluoromethyl)phenylboronic acid pinacol ester ≥97%
4-Ethoxy-2-(trifluoromethyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₀BF₃O₃
MW: 316.124 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24039831
CAS Number: 2121512-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR