2-Benzyloxy-3-chloro-5-fluorophenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

33550-500MG 33550-1G 33550-5G 33550-25G
77325-528EA 128.75 USD
77325-528 77344-238 77361-898 77373-478
2-Benzyloxy-3-chloro-5-fluorophenylboronic acid pinacol ester ≥97%
2-Benzyloxy-3-chloro-5-fluorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₉H₂₁BClFO₃
Storage Temperature: Ambient
MDL Number: MFCD31382553
CAS Number: 2121512-22-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR