Propan-2-yl 3-bromo-5-isopropylbenzoate ≥97%

Supplier: AOBChem USA

Warning

33506-500MG 33506-1G 33506-5G 33506-25G
77325-520EA 145.23 USD
77325-520 77345-124 77361-888 77384-948
Propan-2-yl 3-bromo-5-isopropylbenzoate ≥97%
Propan-2-yl 3-bromo-5-isopropylbenzoate

Precaución: for Research Use Only

Formula: C₁₃H₁₇BrO₂
MW: 285.177 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30834070
CAS Number: 2121515-29-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR