3-Bromo-2-methoxy-5-methylbenzyl alcohol ≥95%

Supplier: AOBChem USA

Warning

33493-500MG 33493-1G 33493-5G 33493-25G
77325-518EA 184.37 USD
77325-518 77347-188 77361-886 77384-946
3-Bromo-2-methoxy-5-methylbenzyl alcohol ≥95%
3-Bromo-2-methoxy-5-methylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₉H₁₁BrO₂
MW: 231.086 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00196830
CAS Number: 86259-91-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR