Methyl 2-(benzyloxy)-4,5-difluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

33486-500MG 33486-1G 33486-5G 33486-25G
77325-516EA 119.48 USD
77325-516 77343-590 77361-884 77373-474
Methyl 2-(benzyloxy)-4,5-difluorobenzoate ≥97%
Methyl 2-(benzyloxy)-4,5-difluorobenzoate

Precaución: for Research Use Only

Formula: C₁₅H₁₂F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD26960221
CAS Number: 1823267-50-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR