Ethyl 3-ethylbenzoate ≥97%

Supplier: AOBChem USA

Warning

33474-500MG 33474-1G 33474-5G 33474-25G
77325-512EA 141.11 USD
77325-512 77344-788 77361-880 77384-944
Ethyl 3-ethylbenzoate ≥97%
Ethyl 3-ethylbenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₂
MW: 178.228 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09997032
CAS Number: 136569-05-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR