5-Chloro-2-isoproxypyridine-4-boronic acid pinacol ester ≥95%

Supplier: AOBChem USA

Warning

33440-500MG 33440-1G 33440-5G 33440-25G
77325-508EA 263.68 USD
77325-508 77349-222 77361-876 77384-940
5-Chloro-2-isoproxypyridine-4-boronic acid pinacol ester ≥95%
5-Chloro-2-isoproxypyridine-4-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₂₁BClNO₃
MW: 297.585 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30834065
CAS Number: 2121513-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR