5-Chloro-2-(propan-2-yloxy)pyridine-4-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

33438-500MG 33438-1G 33438-5G 33438-25G
77325-506EA 204.97 USD
77325-506 77349-220 77361-874 77384-938
5-Chloro-2-(propan-2-yloxy)pyridine-4-carboxylic acid ≥97%
5-Chloro-2-(propan-2-yloxy)pyridine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₁₀ClNO₃
MW: 215.634 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30834063
CAS Number: 1602466-28-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR