Methyl 6-butoxy-5-methylpyridine-3-carboxylate ≥97%

Supplier: AOBChem USA

Warning

33421-500MG 33421-1G 33421-5G
77335-668EA 363.59 USD
77335-668 77349-216 77373-466
Methyl 6-butoxy-5-methylpyridine-3-carboxylate ≥97%
Methyl 6-butoxy-5-methylpyridine-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₇NO₃
MW: 223.268 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30834060
CAS Number: 2121515-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR