3-Bromo-5-ethyl-2-fluorophenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

33416-500MG 33416-1G 33416-5G
77337-740EA 440.84 USD
77337-740 77349-214 77373-464
3-Bromo-5-ethyl-2-fluorophenylboronic acid pinacol ester ≥97%
3-Bromo-5-ethyl-2-fluorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₉BBrFO₂
MW: 329.013 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31382550
CAS Number: 2121514-74-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR