(4-(Benzyloxy)-2,3-dimethylphenyl)boronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

33343-500MG 33343-1G 33343-5G 33343-25G
77325-502EA 119.48 USD
77325-502 77343-588 77361-864 77373-458
(4-(Benzyloxy)-2,3-dimethylphenyl)boronic acid pinacol ester ≥97%
(4-(Benzyloxy)-2,3-dimethylphenyl)boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₂₁H₂₇BO₃
Storage Temperature: Ambient
MDL Number: MFCD30834047
CAS Number: 2121514-48-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR