4-(Benzyloxy)-2,3-dimethylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

33341-500MG 33341-1G 33341-5G 33341-25G
77325-498EA 119.48 USD
77325-498 77343-586 77361-860 77373-456
4-(Benzyloxy)-2,3-dimethylbenzoic acid ≥97%
4-(Benzyloxy)-2,3-dimethylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₆H₁₆O₃
MW: 256.296 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31620766
CAS Number: 148581-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR