5-Ethyl-2-fluorobenzoic acid ≥95%

Supplier: AOBChem USA

Warning

33336-500MG 33336-1G 33336-5G 33336-25G
77325-494EA 222.48 USD
77325-494 77349-206 77361-856 77384-930
5-Ethyl-2-fluorobenzoic acid ≥95%
5-Ethyl-2-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₂
MW: 168.165 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18411936
CAS Number: 1261451-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR