3-(9H-Carbazol-9-yl)-5-chlorophenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

33329-500MG 33329-1G 33329-5G
77339-286EA 572.68 USD
77339-286 77349-204 77373-454
3-(9H-Carbazol-9-yl)-5-chlorophenylboronic acid ≥95%
3-(9H-Carbazol-9-yl)-5-chlorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₈H₁₃BClNO₂
MW: 321.565 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30834040
CAS Number: 2121514-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR