Methyl 5-chloro-2-ethoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

33314-500MG 33314-1G 33314-5G 33314-25G
77325-486EA 244.11 USD
77325-486 77349-200 77361-848 77384-922
Methyl 5-chloro-2-ethoxybenzoate ≥97%
Methyl 5-chloro-2-ethoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO₃
Storage Temperature: Ambient
MDL Number: MFCD16994149
CAS Number: 75230-37-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR