2-Aminopyridine-5-boronic acid pinacol ester hydrochloride ≥95%

Supplier: AOBChem USA

Warning

33294-1G 33294-5G 33294-25G
77341-594EA 94.76 USD
77341-594 77361-846 77373-452
2-Aminopyridine-5-boronic acid pinacol ester hydrochloride ≥95%
2-Aminopyridine-5-boronic acid pinacol ester hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₈BClN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD30737500
CAS Number: 2121514-56-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR