(4-Bromo-3,5-dimethylphenoxy)(tert-butyl)dimethylsilane ≥97%

Supplier: AOBChem USA

Warning

33266-500MG 33266-1G 33266-5G
77337-480EA 415.09 USD
77337-480 77349-198 77373-450
(4-Bromo-3,5-dimethylphenoxy)(tert-butyl)dimethylsilane ≥97%
(4-Bromo-3,5-dimethylphenoxy)(tert-butyl)dimethylsilane

Precaución: for Research Use Only

Formula: C₁₄H₂₃BrOSi
MW: 315.321 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30296244
CAS Number: 149228-92-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR