4-Benzyloxy-3,5-dimethyl-benzoic acid methyl ester ≥95%

Supplier: AOBChem USA

Warning

33260-500MG 33260-1G 33260-5G 33260-25G
77325-482EA 254.41 USD
77325-482 77349-196 77361-842 77384-918
4-Benzyloxy-3,5-dimethyl-benzoic acid methyl ester ≥95%
4-Benzyloxy-3,5-dimethyl-benzoic acid methyl ester

Precaución: for Research Use Only

Formula: C₁₇H₁₈O₃
MW: 270.323 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06203769
CAS Number: 773873-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR