1-Iodo-4-methoxy-2-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

33256-500MG 33256-1G 33256-5G 33256-25G
77325-480EA 99.91 USD
77325-480 77342-774 77361-840 77373-448
1-Iodo-4-methoxy-2-(trifluoromethyl)benzene ≥97%
1-Iodo-4-methoxy-2-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₈H₆F₃IO
MW: 302.032 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08704331
CAS Number: 1261645-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR