Methyl 2-(propan-2-yloxy)pyridine-4-carboxylate ≥97%

Supplier: AOBChem USA

Warning

33241-500MG 33241-1G 33241-5G 33241-25G
77325-478EA 184.37 USD
77325-478 77347-186 77361-838 77384-916
Methyl 2-(propan-2-yloxy)pyridine-4-carboxylate ≥97%
Methyl 2-(propan-2-yloxy)pyridine-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₃
MW: 195.215 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23113734
CAS Number: 262296-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR