5-Bromo-2-hydroxy-4-methylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

33228-1G 33228-5G 33228-25G
77340-892EA 62.83 USD
77340-892 77361-836 77373-446
5-Bromo-2-hydroxy-4-methylbenzoic acid ≥95%
5-Bromo-2-hydroxy-4-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₇BrO₃
Storage Temperature: Ambient
MDL Number: MFCD11044841
CAS Number: 6623-35-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR