Methyl 3,4-difluoro-5-methoxybenzoate ≥95%

Supplier: AOBChem USA

Warning

33182-500MG 33182-1G 33182-5G 33182-25G
77325-476EA 160.68 USD
77325-476 77345-774 77361-834 77384-914
Methyl 3,4-difluoro-5-methoxybenzoate ≥95%
Methyl 3,4-difluoro-5-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₈F₂O₃
MW: 202.155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22490605
CAS Number: 1806332-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR