4,5-Difluoro-3-methoxybezenemethanol ≥97%

Supplier: AOBChem USA

Warning

33180-500MG 33180-1G 33180-5G 33180-25G
77325-474EA 181.28 USD
77325-474 77346-810 77361-832 77384-912
4,5-Difluoro-3-methoxybezenemethanol ≥97%
4,5-Difluoro-3-methoxybezenemethanol

Precaución: for Research Use Only

Formula: C₈H₈F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD04972455
CAS Number: 887585-14-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR